Dipropyl-2,5-pyridinedicarboxylate ≥98.0%

Supplier: TCI America

Synonyms: Dipropyl Isocinchomeronate, 2,5-Pyridinedicarboxylic Acid Dipropyl Ester, Isocinchomeronic Acid Dipropyl Ester

P0835-25G P0835-5G
TCP0835-025GEA 82.63 CAD
TCP0835-025G TCP0835-5G
Dipropyl-2,5-pyridinedicarboxylate ≥98.0%
Dipropyl-2,5-pyridinedicarboxylate

CAS Number: 136-45-8
MDL Number: MFCD00055315
Molecular Formula: C13H17NO4
Molecular Weight: 251.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 175
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: Dipropyl Isocinchomeronate, 2,5-Pyridinedicarboxylic Acid Dipropyl Ester, Isocinchomeronic Acid Dipropyl Ester

Formula: C₁₃H₁₇NO₄
MW: 251.28 g/mol
Boiling Pt: 175 °C
Density: 1.12 (20 °C)
MDL Number: MFCD00055315
CAS Number: 136-45-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b44249204b932bf53ae1fe1c8a7aeb4cce6e495a&vid=b44249204b932bf53ae1fe1c8a7aeb4cce6e495a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR