2-Amino-6-chloropurine riboside ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

A2054-1G A2054-5G
TCA2054-1GEA 72.68 CAD
TCA2054-1G TCA2054-5G
2-Amino-6-chloropurine riboside ≥95.0% (by HPLC, titration analysis)
2-Amino-6-chloropurine riboside

CAS Number: 2004-07-1
MDL Number: MFCD00005735
Molecular Formula: C10H12ClN5O4
Molecular Weight: 301.69
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Melting point (°C): 175
Specific rotation [a]20/D: -38 deg (C=0.1, H2O)


Product Categories: Bioscience, Nucleosides and Their Analogs
Other Product Names / Synonyms: 2-Amino-6-chloro-9-(beta-D-ribofuranosyl)purine

Formula: C₁₀H₁₂ClN₅O₄
Melting Pt: 175 °C
MDL Number: MFCD00005735
CAS Number: 2004-07-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfeaec70854089d7b89711ebf8cf10236c02ecba&vid=dfeaec70854089d7b89711ebf8cf10236c02ecba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR