Phosphocholine chloride sodium salt hydrate ≥98.0%
Supplier: TCI America
Synonyms:
Phosphorylcholine Chloride Sodium Salt
P0834-1G
P0834-5G
TCP0834-001GEA
211.72
CAD
TCP0834-001G
TCP0834-5G
Phosphocholine chloride sodium salt hydrate ≥98.0%
Phosphocholine chloride sodium salt hydrate
[for Cholinesterase Substrate]
CAS Number: 16904-96-4
MDL Number: MFCD00070597
Molecular Formula: C5H15ClNO4P
Molecular Weight: 263.57
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Other Product Names / Synonyms: Phosphorylcholine Chloride Sodium Salt
Formula:
C₅H₁₅ClNO₄P |
MDL Number:
MFCD00070597 CAS Number: 16904-96-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...