N,N,N',N'',N''-Pentakis(2-hydroxypropyl)diethylenetriamine ≥95.0% (by titrimetric analysis)
Supplier: TCI America
P0832-500G
P0832-25G
P0832-100G
TCP0832-500GEA
601.66
CAD
TCP0832-500G
TCP0832-025G
TCP0832-100G
N,N,N',N'',N''-Pentakis(2-hydroxypropyl)diethylenetriamine ≥95.0% (by titrimetric analysis)
N,N,N',N'',N''-Pentakis(2-hydroxypropyl)diethylenetriamine
CAS Number: 17121-34-5
MDL Number: MFCD00012064
Molecular Formula: C19H43N3O5
Molecular Weight: 393.57
Purity/Analysis Method: >95.0% (T)
Form: Clear Liquid
Formula:
C₁₉H₄₃N₃O₅ MW: 393.57 g/mol |
MDL Number:
MFCD00012064 CAS Number: 17121-34-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...