N,N'-1,4-Phenylenedimaleimide ≥98.0%
Supplier: TCI America
Synonyms:
N,N'-1,4-Bismaleimidobenzene, N,N'-1,4-Phenylenebismaleimide
P0778-5G
P0778-1G
TCP0778-005GEA
165.9
CAD
TCP0778-005G
TCP0778-001G
N,N'-1,4-Phenylenedimaleimide ≥98.0%
N,N'-1,4-Phenylenedimaleimide
CAS Number: 3278-31-7
MDL Number: MFCD00005505
Molecular Formula: C14H8N2O4
Molecular Weight: 268.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: N,N'-1,4-Bismaleimidobenzene, N,N'-1,4-Phenylenebismaleimide
Formula:
C₁₄H₈N₂O₄ |
MDL Number:
MFCD00005505 CAS Number: 3278-31-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...