L-O-Phosphoserine ≥98.0%

Supplier: TCI America

Synonyms: H-Ser(PO₃H₂)-OH, H-Ser(PO3H2)-OH

P0773-5G P0773-25G
TCP0773-005GEA 127.59 CAD
TCP0773-005G TCP0773-025G
L-O-Phosphoserine ≥98.0%
L-O-Phosphoserine

CAS Number: 407-41-0
MDL Number: MFCD00065935
Molecular Formula: C3H8NO6P
Molecular Weight: 185.07
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 167
Specific rotation [a]20/D: 5 deg (C=2, H2O)

Formula: C₃H₈NO₆P
MW: 185.07 g/mol
Melting Pt: 167 °C
MDL Number: MFCD00065935
CAS Number: 407-41-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82dda6e2dde40deb5d1f4cf9d429546b6f8695eb&vid=82dda6e2dde40deb5d1f4cf9d429546b6f8695eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR