2-Propylcyclohexanol (cis- and trans- mixture) ≥90.0%
Supplier: TCI America
P0770-25G
P0770-5G
TCP0770-025GEA
281.22
CAD
TCP0770-025G
TCP0770-005G
2-Propylcyclohexanol (cis- and trans- mixture) ≥90.0%
2-Propylcyclohexanol (cis- and trans- mixture)
CAS Number: 90676-25-8
MDL Number: MFCD00019331
Molecular Formula: C9H18O
Molecular Weight: 142.24
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.92
Formula:
C₉H₁₈O Density: 0.92 (20 °C) |
MDL Number:
MFCD00019331 CAS Number: 90676-25-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...