Biphenyl-3-ol ≥97.0% for synthesis

Supplier: TCI America
Warning

Synonyms: 3-Phenylphenol, [1,1'-Biphenyl]-3-ol, 3-Hydroxybiphenyl, 3-Hydroxydiphenyl

P0768-5G P0768-25G
TCP0768-005GEA 220.74 CAD
TCP0768-005G TCP0768-25G
Biphenyl-3-ol ≥97.0% for synthesis
Biphenyl-3-ol

CAS Number: 580-51-8
MDL Number: MFCD00002294
Molecular Formula: C12H10O
Molecular Weight: 170.21
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 78


Other Product Names / Synonyms: Biphenyl-3-ol, 3-Hydroxybiphenyl

Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 334
Melting Pt: 78
MDL Number: MFCD00002294
CAS Number: 580-51-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd1bac67896aa780be5e64f40d36de1da3501f19&vid=fd1bac67896aa780be5e64f40d36de1da3501f19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR