4-(3-Phenylpropyl)piperidine ≥98.0%

Supplier: TCI America

P0760-25G P0760-5G
TCP0760-25GEA 222.78 CAD
TCP0760-25G TCP0760-5G
4-(3-Phenylpropyl)piperidine ≥98.0%
4-(3-Phenylpropyl)piperidine

CAS Number: 18495-82-4
MDL Number: MFCD00023152
Molecular Formula: C14H21N
Molecular Weight: 203.33
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 0.96

Formula: C₁₄H₂₁N
MW: 203.33 g/mol
Density: 0.96 (20 °C)
MDL Number: MFCD00023152
CAS Number: 18495-82-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98d0680fa8ac606abbc4f628691a38c82af41d9d&vid=98d0680fa8ac606abbc4f628691a38c82af41d9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR