3'-Azido-3'-deoxythymidine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A2052-5G A2052-1G
TCA2052-5GEA 216.46 CAD
TCA2052-5G TCA2052-1G
3'-Azido-3'-deoxythymidine ≥98.0% (by titrimetric analysis)
3'-Azido-3'-deoxythymidine

CAS Number: 30516-87-1
MDL Number: MFCD00006536
Molecular Formula: C10H13N5O4
Molecular Weight: 267.25
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 124
Specific rotation [a]20/D: 47 deg (C=1, H2O)


Product Categories: Synthetic Organic Chemistry, Azide Compounds (Click Chemistry), Bioscience, Regenerative Medicine Research Reagents, Antiviral Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analo
Other Product Names / Synonyms: Azidothymidine, AZT, 1-(3-Azido-2,3-dideoxy-beta-D-ribofuranosyl)thymine, Zidovudine

Formula: C₁₀H₁₃N₅O₄
Melting Pt: 124 °C
MDL Number: MFCD00006536
CAS Number: 30516-87-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f73272f58701a65c0d1d22d7b10e955daa0afd80&vid=f73272f58701a65c0d1d22d7b10e955daa0afd80&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR