Pentaerythritol tetraacetate ≥98.0%

Supplier: TCI America

P0757-25G P0757-500G
TCP0757-025GEA 109.02 CAD
TCP0757-025G TCP0757-500G
Pentaerythritol tetraacetate ≥98.0%
Pentaerythritol tetraacetate

CAS Number: 597-71-7
MDL Number: MFCD00026206
Molecular Formula: C13H20O8
Molecular Weight: 304.30
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 325
Melting point (°C): 80

Formula: C₁₃H₂₀O₈
Boiling Pt: 325 °C
Melting Pt: 80 °C
MDL Number: MFCD00026206
CAS Number: 597-71-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4078c8d364ff8a0ddea62f853303d97899f3ea66&vid=4078c8d364ff8a0ddea62f853303d97899f3ea66&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR