trans-2-Phenylcyclopropanecarboxylic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

P0750-25G P0750-5G
TCP0750-25GEA 750.5 CAD
TCP0750-25G TCP0750-005G
trans-2-Phenylcyclopropanecarboxylic acid ≥98.0% (by titrimetric analysis)
trans-2-Phenylcyclopropanecarboxylic acid

CAS Number: 939-90-2
MDL Number: MFCD00001292
Molecular Formula: C10H10O2
Molecular Weight: 162.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 88

Formula: C₁₀H₁₀O₂
Melting Pt: 88 °C
MDL Number: MFCD00001292
CAS Number: 939-90-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f196601be00ba934b3477f52da39c6327a2d2617&vid=f196601be00ba934b3477f52da39c6327a2d2617&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR