Pentaerythritol tetrastearate ≥95.0% (by titrimetric analysis) (so called)

Supplier: TCI America

P0739-25G P0739-500G
TCP0739-025GEA 59.42 CAD
TCP0739-025G TCP0739-500G
Pentaerythritol tetrastearate ≥95.0% (by titrimetric analysis) (so called)
Pentaerythritol tetrastearate

CAS Number: 115-83-3
MDL Number: MFCD00041922
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 66
Flash Point (°C): 292

Formula: C₇₇H₁₄₈O₈
MW: 1201.99 g/mol
Melting Pt: 66 °C
Flash Pt: 292 °C
MDL Number: MFCD00041922
CAS Number: 115-83-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=966d9f11a9714d46886e9479c7afcf807a0988be&vid=966d9f11a9714d46886e9479c7afcf807a0988be&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR