9-Acetyl-3,6-diiodocarbazole ≥96.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: 1-(3,6-Diiodo-9H-carbazol-9-yl)ethanone

A2051-1G A2051-5G
TCA2051-1GEA 89.27 CAD
TCA2051-1G TCA2051-5G
9-Acetyl-3,6-diiodocarbazole ≥96.0% (by total nitrogen basis)
9-Acetyl-3,6-diiodocarbazole

CAS Number: 606129-89-9
Molecular Formula: C14H9I2NO
Molecular Weight: 461.04
Purity/Analysis Method: >96.0% (N)
Form: Crystal
Melting point (°C): 228

Formula: C₁₄H₉I₂NO
MW: 461.04 g/mol
Melting Pt: 228 °C
MDL Number: MFCD10000947
CAS Number: 606129-89-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=771a735814711e98b2f800e44646ac7cfd886166&vid=771a735814711e98b2f800e44646ac7cfd886166&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR