Pentyltriethoxysilane ≥97.0%

Supplier: TCI America

P0736-25ML P0736-5ML
TCP0736-025MLEA 618.25 CAD
TCP0736-025ML TCP0736-005ML
Pentyltriethoxysilane ≥97.0%
Pentyltriethoxysilane

CAS Number: 2761-24-2
MDL Number: MFCD00026757
Molecular Formula: C11H26O3Si
Molecular Weight: 234.41
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 205
Flash Point (°C): 83
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: Amyltriethoxysilane, Triethoxypentylsilane

Formula: C₁₁H₂₆O₃Si
Boiling Pt: 205 °C
Density: 0.89 (20 °C)
Flash Pt: 83 °C
MDL Number: MFCD00026757
CAS Number: 2761-24-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25d354cce3b1247fe3e80bfd5178e8ba5f40626d&vid=25d354cce3b1247fe3e80bfd5178e8ba5f40626d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR