Phenylthiohydantoin-proline ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
P0729-100MG
P0729-1G
TCP0729-100MGEA
90.85
CAD
TCP0729-100MG
TCP0729-001G
Phenylthiohydantoin-proline ≥98.0% (by HPLC, total nitrogen)
Phenylthiohydantoin-proline
CAS Number: 4333-21-5
MDL Number: MFCD00056895
Molecular Formula: C12H12N2OS
Molecular Weight: 232.30
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 179
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: PTH-proline
Formula:
C₁₂H₁₂N₂OS Melting Pt: 179 °C |
MDL Number:
MFCD00056895 CAS Number: 4333-21-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...