Polyethylene glycol stearylamine, M.W. n=∼15

Supplier: TCI America

P0725-25G P0725-500G
TCP0725-025GEA 55.3 CAD
TCP0725-025G TCP0725-500G
Polyethylene glycol stearylamine, M.W. n=∼15
Polyethylene glycol stearylamine

MDL Number: MFCD00148008
Form: Clear Liquid
Color: Yellow
Flash Point (°C): 258
Specific Gravity (20/20): 1.01

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR