Polyethylene glycol monostearate n=∼55

Supplier: TCI America

P0723-25G P0723-500G
TCP0723-025GEA 31.6 CAD
TCP0723-025G TCP0723-500G
Polyethylene glycol monostearate n=∼55
Polyethylene glycol monostearate

Palmitate and stearate mixture


Form: Crystal
Color: White

Formula: HO(CH₂CH₂O)nOCC₁₇H₃₅
Melting Pt: 53 °C
MDL Number: MFCD00148007
CAS Number: 9004-99-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR