Polyethylene glycol monostearate n=∼45

Supplier: TCI America

Synonyms: Polyoxyethylene stearate, Polyoxyl stearate

P0722-25G P0722-500G
TCP0722-025GEA 34.76 CAD
TCP0722-025G TCP0722-500G
Polyethylene glycol monostearate n=∼45
Polyethylene glycol monostearate

Palmitate and stearate mixture


Form: Crystal
Color: White

Formula: HO(CH₂CH₂O)nOCC₁₇H₃₅
Melting Pt: 52 °C
MDL Number: MFCD00148007
CAS Number: 9004-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR