Polyethylene glycol monostearate n=∼40
Supplier: TCI America
P0721-25G
P0721-500G
TCP0721-025GEA
34.76
CAD
TCP0721-025G
TCP0721-500G
Polyethylene glycol monostearate n=∼40
Polyethylene glycol monostearate
Palmitate and stearate mixture
Form: Crystal
Color: White
Formula:
HO(CH₂CH₂O)nOCC₁₇H₃₅ Melting Pt: 47 °C Flash Pt: 320 °C |
MDL Number:
MFCD00148007 CAS Number: 9004-99-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...