Polyethylene glycol monostearate n=∼25

Supplier: TCI America

P0720-25G P0720-500G
TCP0720-025GEA 33.76 CAD
TCP0720-025G TCP0720-500G
Polyethylene glycol monostearate n=∼25
Polyethylene glycol monostearate

Palmitate and stearate mixture


Form: Crystal
Color: White

Formula: HO(CH₂CH₂O)nOCC₁₇H₃₅
MDL Number: MFCD00148007
CAS Number: 9004-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR