Polyethylene glycol monostearate n=∼10
Supplier: TCI America
P0719-500G
P0719-25G
TCP0719-500GEA
135.88
CAD
TCP0719-500G
TCP0719-025G
Polyethylene glycol monostearate n=∼10
Polyethylene glycol monostearate
Palmitate and stearate mixture
Form: Crystal
Color: White
Formula:
HO(CH₂CH₂O)nOCC₁₇H₃₅ Flash Pt: 266 °C |
MDL Number:
MFCD00148007 CAS Number: 9004-99-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...