Polyethylene glycol monostearate n=∼4
Supplier: TCI America
Synonyms:
Polyoxyethylene stearate, Polyoxyl stearate
P0718-25G
P0718-500G
TCP0718-025GEA
33.76
CAD
TCP0718-025G
TCP0718-500G
Polyethylene glycol monostearate n=∼4
Polyethylene glycol monostearate
Palmitate and stearate mixture
Form: Crystal
Color: White
Formula:
HO(CH₂CH₂O)nOCC₁₇H₃₅ |
MDL Number:
MFCD00148007 CAS Number: 9004-99-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...