Polyethylene glycol monooleyl ether, M.W. n=∼50
Supplier: TCI America
P0716-25G
P0716-500G
TCP0716-025GEA
26.86
CAD
TCP0716-025G
TCP0716-500G
Polyethylene glycol monooleyl ether, M.W. n=∼50
Polyethylene glycol monooleyl ether
CAS Number: 9004-98-2
MDL Number: MFCD00074369
Form: Flake
Color: White
Flash Point (°C): 150
Product Categories: Materials Chemistry, Nonionic Surfactants
Formula:
HO(CH₂CH₂O)nC₁₈H₃₅ Flash Pt: 150 °C |
MDL Number:
MFCD00074369 CAS Number: 9004-98-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...