Polyethylene glycol monooleyl ether, M.W. n=∼20

Supplier: TCI America

P0715-25G P0715-500G
TCP0715-025GEA 30.02 CAD
TCP0715-025G TCP0715-500G
Polyethylene glycol monooleyl ether, M.W. n=∼20
Polyethylene glycol monooleyl ether

CAS Number: 9004-98-2
MDL Number: MFCD00074369
Form: Crystal
Color: White
Melting point (°C): 35
Flash Point (°C): 150


Product Categories: Materials Chemistry, Nonionic Surfactants

Formula: HO(CH₂CH₂O)nC₁₈H₃₅
Melting Pt: 35 °C
Flash Pt: 150 °C
MDL Number: MFCD00074369
CAS Number: 9004-98-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=185b0a46186508563b024440f7cb9e7126bd0337&vid=4281d98cdafd10839871eada56feef6103a40d3e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR