Polyethylene glycol monooleyl ether, M.W. n=∼10
Supplier: TCI America
P0714-25G
P0714-500G
TCP0714-025GEA
30.02
CAD
TCP0714-025G
TCP0714-500G
Polyethylene glycol monooleyl ether, M.W. n=∼10
Polyethylene glycol monooleyl ether
CAS Number: 9004-98-2
MDL Number: MFCD00074369
Form: Paste
Color: Very Pale Yellow
Flash Point (°C): 251
Product Categories: Materials Chemistry, Nonionic Surfactants
Formula:
HO(CH2CH2O)nC18H35 Flash Pt: 251 °C |
MDL Number:
MFCD00074369 CAS Number: 9004-98-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...