3-Aminopyridazine ≥98.0%

Supplier: TCI America

Synonyms: Pyridazin-3-amine

A2048-1G A2048-5G
TCA2048-1GEA 41.08 CAD
TCA2048-1G TCA2048-5G
3-Aminopyridazine ≥98.0%
3-Aminopyridazine

CAS Number: 5469-70-5
MDL Number: MFCD01529869
Molecular Formula: C4H5N3
Molecular Weight: 95.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 171

Formula: C4H5N3
MW: 95.10 g/mol
Melting Pt: 171 °C
MDL Number: MFCD01529869
CAS Number: 5469-70-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=426f30645bc3e12d453396296110eb7af390df49&vid=426f30645bc3e12d453396296110eb7af390df49&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR