2-Phenylimidazoline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

P0685-500G P0685-25G
TCP0685-500GEA 413.17 CAD
TCP0685-500G TCP0685-25G
2-Phenylimidazoline ≥98.0% (by GC, titration analysis)
2-Phenylimidazoline

CAS Number: 936-49-2
MDL Number: MFCD00005180
Molecular Formula: C9H10N2
Molecular Weight: 146.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 181
Melting point (°C): 100
Lambda max.: 325 nm

Formula: C₉H₁₀N₂
Boiling Pt: 181 °C
Melting Pt: 100 °C
MDL Number: MFCD00005180
CAS Number: 936-49-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd953baf58d5dbbcd73b0434e44ecf58db399d39&vid=bd953baf58d5dbbcd73b0434e44ecf58db399d39&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR