Tris(2-butoxyethyl)phosphate ≥95.0%

Supplier: TCI America

P0683-25G P0683-500G
TCP0683-025GEA 54.51 CAD
TCP0683-025G TCP0683-500G
Tris(2-butoxyethyl)phosphate ≥95.0%
Tris(2-butoxyethyl)phosphate

CAS Number: 78-51-3
MDL Number: MFCD00009456
Molecular Formula: C18H39O7P
Molecular Weight: 398.48
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 220
Flash Point (°C): 210
Specific Gravity (20/20): 1.02


Product Categories: Materials Chemistry, Secondary Battery Research Reagents, Resin Additives
Other Product Names / Synonyms: Phosphoric Acid Tris(2-butoxyethyl) Ester

Formula: C₁₈H₃₉O₇P
Boiling Pt: 220 °C
Density: 1.02 (20 °C)
Flash Pt: 210 °C
MDL Number: MFCD00009456
CAS Number: 78-51-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c6229fdd0ad767d434fc1a4ea73234a0f7b1470&vid=0c6229fdd0ad767d434fc1a4ea73234a0f7b1470&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR