Aminomethyl polystyrene resin 2.0-3.0 mmol/g, cross-linked with 1% DVB (200-400mesh)
Supplier: TCI America
A2047-25G A2047-5G
TCA2047-25GEA 901.66 CAD
TCA2047-25G TCA2047-5G
Aminomethyl polystyrene resin 2.0-3.0 mmol/g, cross-linked with 1% DVB (200-400mesh)
Aminomethyl polystyrene resin
Color: Very Pale Yellow
Product Categories: Synthetic Organic Chemistry, Resins (Combinatorial Chemistry)
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31b71be551d745ce9577f211e0ec90f522fc1a42&vid=b1cfe301f558e704143fbccbc7d9603b05359524&prid=web&clid=SAPDAM