Poly(N-vinylcarbazole)
Supplier: TCI America
P0656-25G
P0656-5G
TCP0656-25GEA
948
CAD
TCP0656-25G
TCP0656-5G
Poly(N-vinylcarbazole)
Poly(N-vinylcarbazole)
CAS Number: 25067-59-8
MDL Number: MFCD00134336
Form: Crystal
Color: White
Other Product Names / Synonyms: N-Vinylcarbazole Polymer
MDL Number:
MFCD00134336 CAS Number: 25067-59-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...