4-Amino-1-boc-piperidine ≥97.0%
Supplier: TCI America
Synonyms:
tert-Butyl 4-Amino-1-piperidinecarboxylate, 4-Amino-1-tert-butoxycarbonylpiperidine, tert-Butyl 4-aminopiperidine-1-carboxylate, N-Boc-4-piperidineamine, tert-Butyl 4-amino-1-piperidinecarboxylate, 4-Amino-1-Boc-piperidine, 4-Amino-1-piperidinecarboxylic acid tert-Butyl Ester
CAS Number: 87120-72-7
MDL Number: MFCD01076201
Molecular Formula: C10H20N2O2
Molecular Weight: 200.28
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 45
Other Product Names / Synonyms: 4-Amino-1-Boc-piperidine, tert-Butyl 4-Amino-1-piperidinecarboxylate, 4-Amino-1-piperidinecarboxylic Acid tert-Butyl Ester
Formula:
C₁₀H₂₀N₂O₂ MW: 200.28 g/mol Melting Pt: 45 °C |
MDL Number:
MFCD01076201 CAS Number: 87120-72-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...