Phenylthiohydantoin(Nε-phenylthiocarbamyl)lysine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

P0625-100MG P0625-1G
TCP0625-100MGEA 45.82 CAD
TCP0625-100MG TCP0625-001G
Phenylthiohydantoin(Nε-phenylthiocarbamyl)lysine ≥98.0% (by HPLC, titration analysis)
Phenylthiohydantoin(Nepsilon-phenylthiocarbamyl)lysine

CAS Number: 29635-94-7
MDL Number: MFCD00151476
Molecular Formula: C20H22N4OS2
Molecular Weight: 398.54
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 165


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: PTH-(Nepsilon-PTC)-lysine

Formula: C₂₀H₂₂N₄OS₂
Melting Pt: 165 °C
MDL Number: MFCD00151476
CAS Number: 29635-94-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8db74ec007d0822428ef9fe8f818723a637e39a&vid=f8db74ec007d0822428ef9fe8f818723a637e39a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR