Pontacyl carmine 2B ≥93.0% (by HPLC)

Supplier: TCI America

P0595-500G P0595-25G
TCP0595-500GEA 656.17 CAD
TCP0595-500G TCP0595-025G
Pontacyl carmine 2B ≥93.0% (by HPLC)
Pontacyl carmine 2B

CAS Number: 6625-46-3
MDL Number: MFCD00059131
Molecular Formula: C19H17N3O9S2
Molecular Weight: 539.44
Purity/Analysis Method: >93.0% (HPLC)
Form: Crystal

Formula: C₁₉H₁₇N₃O₉S₂
MDL Number: MFCD00059131
CAS Number: 6625-46-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=440729ad72fb43aaac3960b32cccaf6d33aed7cb&vid=440729ad72fb43aaac3960b32cccaf6d33aed7cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (HPLC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR