1-(4-Pyridyl)pyridinium chloride hydrochloride hydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: [1,4-Bipyridin]-1-ium chloride hydrochloride

P0565-25G P0565-5G
TCP0565-025GEA 237 CAD
TCP0565-025G 76220-196
1-(4-Pyridyl)pyridinium chloride hydrochloride hydrate ≥98.0% (by HPLC, titration analysis)
1-(4-Pyridyl)pyridinium chloride hydrochloride hydrate

CAS Number: 5421-92-1
MDL Number: MFCD00012821
Molecular Formula: C10H9ClN2
Molecular Weight: 229.10
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Formula: C₁₀H₉ClN₂·HCl·xH₂O
MW: 229.10 g/mol (anhy)
MDL Number: MFCD00012821
CAS Number: 5421-92-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d048f83af72577f40914f9ddbe33b3b4e1c34e62&vid=79c194b9286eb9eb5381fcb45d910391802d8f6b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR