8-Hydroxy-7-propylquinoline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

P0531-5G P0531-1G
TCP0531-005GEA 1153.72 CAD
TCP0531-005G TCP0531-1G
8-Hydroxy-7-propylquinoline ≥98.0% (by GC, titration analysis)
8-Hydroxy-7-propylquinoline

CAS Number: 58327-60-9
MDL Number: MFCD00059755
Molecular Formula: C12H13NO
Molecular Weight: 187.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 149
Melting point (°C): 0
Specific Gravity (20/20): 1.10


Other Product Names / Synonyms: 7-Propyloxine, 7-Propyl-8-quinolinol

Formula: C₁₂H₁₃NO
Boiling Pt: 149 °C
Density: 1.10 (20 °C)
MDL Number: MFCD00059755
CAS Number: 58327-60-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52c5fcfd49a49cf18c573740ff376976673e6fb6&vid=52c5fcfd49a49cf18c573740ff376976673e6fb6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR