Tetrahydro-2-pyrimidinone ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Tetrahydro-2(1H)-pyrimidinone, N,N'-Trimethyleneurea, Propyleneurea

P0526-25G P0526-500G
TCP0526-025GEA 109.02 CAD
TCP0526-025G TCP0526-500G
Tetrahydro-2-pyrimidinone ≥97.0% (by titrimetric analysis)
Tetrahydro-2-pyrimidinone

CAS Number: 1852-17-1
MDL Number: MFCD00014593
Molecular Formula: C4H8N2O
Molecular Weight: 100.12
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 266
Flash Point (°C): 158


Other Product Names / Synonyms: Propyleneurea, N,N'-Trimethyleneurea

Formula: C₄H₈N₂O
MW: 100.12 g/mol
Melting Pt: 266 °C
Flash Pt: 158 °C
MDL Number: MFCD00014593
CAS Number: 1852-17-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8628152c7f9a34147d0eb5fe6b06173189bd9ef3&vid=8628152c7f9a34147d0eb5fe6b06173189bd9ef3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR