D-Arginine methyl ester dihydrochloride ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

A2016-1G A2016-5G
TCA2016-1GEA 97.96 CAD
TCA2016-1G TCA2016-5G
D-Arginine methyl ester dihydrochloride ≥98.0% (by total nitrogen and titration analysis)
D-Arginine methyl ester dihydrochloride

CAS Number: 78851-84-0
MDL Number: MFCD00153417
Molecular Formula: C7H16N4O2
Molecular Weight: 261.15
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Specific rotation [a]20/D: -21 deg (C=2.5, MeOH)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-D-Arg-OMe.2HCl

Formula: C₇H₁₆N₄O₂
MDL Number: MFCD00153417
CAS Number: 78851-84-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0f4c0660b2006d7448049c2740948be00619b9a&vid=e0f4c0660b2006d7448049c2740948be00619b9a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR