Piperazine dihydrochloride monohydrate ≥98.0%

Supplier: TCI America

P0450-25G P0450-500G
TCP0450-025GEA 53.06 CAD
TCP0450-025G TCP0450-500G
Piperazine dihydrochloride monohydrate ≥98.0%
Piperazine dihydrochloride monohydrate

CAS Number: 142-64-3
MDL Number: MFCD00012754
Molecular Formula: C4H10N2
Molecular Weight: 159.05
Purity/Analysis Method: >98.0% (N)
Form: Crystal

Formula: C₄H₁₀N₂·2HCl·H₂O
MW: 177.07 g/mol
MDL Number: MFCD00012754
CAS Number: 6091-62-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57c1758d5192eb4e5faada1844dd8a4e298baa0d&vid=57c1758d5192eb4e5faada1844dd8a4e298baa0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR