Piperazine citrate hydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
P0449-25G
P0449-500G
TCP0449-025GEA
50.56
CAD
TCP0449-025G
TCP0449-500G
Piperazine citrate hydrate ≥98.0% (by titrimetric analysis)
Piperazine citrate hydrate
CAS Number: 144-29-6
MDL Number: MFCD00038961
Molecular Formula: C4H10N2
Molecular Weight: 642.66
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 185
Formula:
C₄H₁₀N₂ Melting Pt: 185 °C |
MDL Number:
MFCD00038961 CAS Number: 144-29-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...