Piperazine adipate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

P0448-25G P0448-500G
TCP0448-025GEA 45.82 CAD
TCP0448-025G TCP0448-500G
Piperazine adipate ≥98.0% (by titrimetric analysis)
Piperazine adipate

CAS Number: 142-88-1
MDL Number: MFCD00050650
Molecular Formula: C4H10N2
Molecular Weight: 232.28
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₄H₁₀N₂
MDL Number: MFCD00050650
CAS Number: 142-88-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae85a7d33ad2970b2831804e06e4e9065ed04624&vid=ae85a7d33ad2970b2831804e06e4e9065ed04624&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR