D-Alanine methyl ester hydrochloride ≥97.0%

Supplier: TCI America

Synonyms: D-Alanine Methyl Ester Hydrochloride, (R)-2-Aminopropionic acid Methyl Ester Hydrochloride, H-D-Ala-OMe.HCl, H-D-Ala-OMe·HCl, Methyl D-alaninate hydrochloride, Methyl (R)-2-Aminopropionate Hydrochloride, (R)-2-Aminopropionic Acid Methyl Ester Hydrochloride, (R)-Methyl 2-aminopropionate hydrochloride

A2011-25G A2011-5G
TCA2011-25GEA 170.32 CAD
TCA2011-25G TCA2011-5G
D-Alanine methyl ester hydrochloride ≥97.0%
D-Alanine methyl ester hydrochloride

CAS Number: 14316-06-4
MDL Number: MFCD00066141
Molecular Formula: C4H9NO2
Molecular Weight: 139.58
Purity/Analysis Method: >97.0% (N,T)
Form: Crystal
Melting point (°C): 108
Specific rotation [a]20/D: -8 deg (C=2, MeOH)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: Methyl (R)-2-Aminopropionate Hydrochloride, (R)-2-Aminopropionic Acid Methyl Ester Hydrochloride, H-D-Ala-OMe.HCl

Formula: C₄H₉NO₂
MW: 139.58 g/mol
Melting Pt: 108 °C
MDL Number: MFCD00066141
CAS Number: 14316-06-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=000f7f740cf489df492cd8340a42d4c20561c0a3&vid=000f7f740cf489df492cd8340a42d4c20561c0a3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR