Picolinic acid hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: Pyridine-2-carboxylic acid hydrochloride, Picolinic acid hydrochloride

P0422-25G P0422-5G
TCP0422-025GEA 352.02 CAD
TCP0422-025G TCP0422-5G
Picolinic acid hydrochloride ≥98.0%
Picolinic acid hydrochloride

CAS Number: 636-80-6
MDL Number: MFCD00050666
Molecular Formula: C6H5NO2
Molecular Weight: 159.57
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: Picolinic Acid Hydrochloride

Formula: C₆H₆ClNO₂
MW: 159.5703 g/mol
MDL Number: MFCD00050666
CAS Number: 636-80-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e7d3b1e0a8594af63f78117094d88e232c5cb55&vid=1e7d3b1e0a8594af63f78117094d88e232c5cb55&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR