Phytic acid ∼50% ∼1.1 mol/L in water
Supplier: TCI America
Synonyms:
Inositol hexaphosphoric acid
P0409-500G
P0409-25G
TCP0409-500GEA
141.92
CAD
TCP0409-500G
TCP0409-025G
Phytic acid ∼50% ∼1.1 mol/L in water
Phytic acid
CAS Number: 83-86-3
MDL Number: MFCD00082309
Molecular Formula: C6H18O24P6
Molecular Weight: 660.03
Form: Clear Liquid
Color: Very Pale Yellow
Storage Temperature: 0-10°C
Formula:
C₆H₁₈O₂₄P₆ MW: 660.03 g/mol |
MDL Number:
MFCD00082309 CAS Number: 83-86-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...