Disodium phthalate ≥95.0% (by titrimetric analysis)
Supplier: TCI America
P0400-25G
P0400-500G
TCP0400-025GEA
55.3
CAD
TCP0400-025G
TCP0400-500G
Disodium phthalate ≥95.0% (by titrimetric analysis)
Disodium phthalate
CAS Number: 15968-01-1
MDL Number: MFCD00050736
Molecular Formula: C8H6O4
Molecular Weight: 210.10
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Other Product Names / Synonyms: Phthalic Acid Disodium Salt
Formula:
C₈H₆O₄ |
MDL Number:
MFCD00050736 CAS Number: 15968-01-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...