L-Alanine-tert-butyl ester hydrochloride ≥97.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: L-Alanine tert-Butyl Ester Hydrochloride, tert-Butyl (S)-2-Aminopropionate Hydrochloride, H-Ala-OtBu·HCl, (S)-2-Aminopropionic acid tert-Butyl Ester Hydrochloride, H-Ala-OtBu.HCl, (S)-2-Aminopropionic Acid tert-Butyl Ester Hydrochloride

A2010-1G A2010-5G
TCA2010-1GEA 88.48 CAD
TCA2010-1G TCA2010-5G
L-Alanine-tert-butyl ester hydrochloride ≥97.0% (by total nitrogen basis)
L-Alanine-tert-butyl ester hydrochloride

CAS Number: 13404-22-3
MDL Number: MFCD00035524
Molecular Formula: C7H15NO2
Molecular Weight: 181.66
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Melting point (°C): 208
Specific rotation [a]20/D: 2 deg (C=2, EtOH)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: tert-Butyl (S)-2-Aminopropionate Hydrochloride, (S)-2-Aminopropionic Acid tert-Butyl Ester Hydrochloride, H-Ala-OtBu.HCl

Formula: C₇H₁₅NO₂
MW: 181.66 g/mol
Melting Pt: 208 °C
MDL Number: MFCD00035524
CAS Number: 13404-22-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=981c461a4db7fa14c7d410b3893412cbacca8bcb&vid=981c461a4db7fa14c7d410b3893412cbacca8bcb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR