Trioleyl phosphite ≥98.0% (by titrimetric analysis) (so called)

Supplier: TCI America

P0390-500ML P0390-25ML
TCP0390-500MLEA 259.12 CAD
TCP0390-500ML TCP0390-025ML
Trioleyl phosphite ≥98.0% (by titrimetric analysis) (so called)
Trioleyl phosphite

CAS Number: 13023-13-7
MDL Number: MFCD00059452
Molecular Formula: C54H105O3P
Molecular Weight: 833.40
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Flash Point (°C): 225
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: Phosphorous Acid Trioleyl Ester

Formula: C₅₄H₁₀₅O₃P
Density: 0.88 (20 °C)
Flash Pt: 225 °C
MDL Number: MFCD00059452
CAS Number: 13023-13-7

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR