Phenylthiohydantoin-tryptophan ≥98.0%

Supplier: TCI America

Synonyms: 5-(3-Indolylmethyl)-3-phenyl-2-thiohydantoin, PTH-tryptophan

P0378-100MG P0378-1G
TCP0378-100MGEA 102.83 CAD
TCP0378-100MG TCP0378-001G
Phenylthiohydantoin-tryptophan ≥98.0%
Phenylthiohydantoin-tryptophan

CAS Number: 5789-24-2
MDL Number: MFCD06858416
Molecular Formula: C18H15N3OS
Molecular Weight: 321.40
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 183


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 5-(3-Indolylmethyl)-3-phenyl-2-thiohydantoin, PTH-tryptophan

Formula: C₁₈H₁₅N₃OS
MW: 321.40 g/mol
Melting Pt: 183 °C
MDL Number: MFCD06858416
CAS Number: 5789-24-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=073947fc46181c1c0965f250e3132f4517cc77ee&vid=073947fc46181c1c0965f250e3132f4517cc77ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR