Phenylthiohydantoin-tyrosine ≥98.0% (by HPLC)

Supplier: TCI America

P0368-1G P0368-100MG
TCP0368-001GEA 281.24 CAD
TCP0368-001G TCP0368-100MG
Phenylthiohydantoin-tyrosine ≥98.0% (by HPLC)
Phenylthiohydantoin-tyrosine

CAS Number: 4332-95-0
MDL Number: MFCD00027415
Molecular Formula: C16H14N2O2S
Molecular Weight: 298.36
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 221


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 5-(4-Hydroxybenzyl)-3-phenyl-2-thiohydantoin, PTH-tyrosine

Formula: C₁₆H₁₄N₂O₂S
Melting Pt: 221 °C
MDL Number: MFCD00027415
CAS Number: 4332-95-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1ee40d94a6a89d6f4f1b570e08b0977424b7c35&vid=a1ee40d94a6a89d6f4f1b570e08b0977424b7c35&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR