Phenylthiohydantoin-phenylalanine

Supplier: TCI America

Synonyms: 5-Benzyl-3-phenyl-2-thiohydantoin, PTH-phenylalanine

P0367-100MG P0367-1G
TCP0367-100MGEA 64.78 CAD
TCP0367-100MG TCP0367-001G
Phenylthiohydantoin-phenylalanine
Phenylthiohydantoin-phenylalanine

CAS Number: 4332-97-2
MDL Number: MFCD01076623
Molecular Formula: C16H14N2OS
Molecular Weight: 282.36
Form: Crystal
Color: White
Melting point (°C): 187


Other Product Names / Synonyms: 5-Benzyl-3-phenyl-2-thiohydantoin, PTH-phenylalanine

Formula: C₁₆H₁₄N₂OS
Melting Pt: 187 °C
MDL Number: MFCD01076623
CAS Number: 4332-97-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66b44d683302489f793bba8a4055aff79792d89c&vid=66b44d683302489f793bba8a4055aff79792d89c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR